Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-5-hydroxyisophthalate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 5-Hydroxyisophthalic acid dimethyl esterDimethyl 5-hydroxyisophthalate

H1007-250G H1007-25G
TCH1007-25GEA 61.32 USD InStock
TCH1007-25G TCH1007-250G
Dimethyl-5-hydroxyisophthalate ≥98.0% (by GC, titration analysis)
Dimethyl 5-hydroxyisophthalate
CAS Number: 13036-02-7
MDL Number: MFCD00134367
Molecular Formula: C10H10O5
Molecular Weight: 210.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 165

Other Product Names / Synonyms: 5-Hydroxyisophthalic Acid Dimethyl Ester


Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00134367
CAS Number: 13036-02-7
EINECS: 235-899-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/10/30/28971030.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty